4-Amino-2,6-dichloropyridine, 98%, Thermo Scientific Chemicals
4-Amino-2,6-dichloropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-2,6-dichloropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 398700050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-amino-2,6-dichloropyridin-1-ium
Molecular FormulaC5H5Cl2N2
InChI KeyWAEZOSSWRXDWAX-UHFFFAOYSA-O
SMILESNC1=CC(Cl)=[NH+]C(Cl)=C1
Molecular Weight (g/mol)164.01
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow-brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point169°C to 173°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only