Thermo Scientific Chemicals

Azetidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 398730050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Azetidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 398730050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 398730050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
36520-39-5
IUPAC Name
azetidine hydrochloride
Molecular Formula
C3H8ClN
InChI Key
HGQULGDOROIPJN-UHFFFAOYSA-N
SMILES
Cl.C1CNC1
Appearance (Color)
White to brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text