Thermo Scientific Chemicals

Ethyl 2-chloro-2-(hydroxyimino)acetate, 97%, Thermo Scientific Chemicals

Catalog number: 398740050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 2-chloro-2-(hydroxyimino)acetate, 97%, Thermo Scientific Chemicals

Catalog number: 398740050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 398740050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14337-43-0
IUPAC Name
ethyl (2Z)-2-chloro-2-(hydroxyimino)acetate
Molecular Formula
C4H6ClNO3
InChI Key
UXOLDCOJRAMLTQ-UTCJRWHESA-N
SMILES
CCOC(=O)C(Cl)=NO
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
70°C to 75°C
Titration Argentometric
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text