Thermo Scientific Chemicals

Tetrakis(acetonitrile)copper(I) tetrafluoroborate, Thermo Scientific Chemicals

Catalog number: 398760010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrakis(acetonitrile)copper(I) tetrafluoroborate, Thermo Scientific Chemicals

Catalog number: 398760010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 398760010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15418-29-8
IUPAC Name
λ¹-copper(1+) tetrakis(acetonitrile) tetrafluoroboranuide
Molecular Formula
C8H12BCuF4N4
InChI Key
YZGSKMIIVMCEFE-UHFFFAOYSA-N
SMILES
[Cu+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration Complexometric
19.5 to 20.5 % Cu

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text