Chemical Identifiers
CAS2627-69-2
IUPAC Name5-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-imidazole-4-carboxamide
Molecular FormulaC9H14N4O5
InChI KeyRTRQQBHATOEIAF-UUOKFMHZSA-N
SMILESNC(=O)C1=C(N)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
View more