Thermo Scientific Chemicals

2-(BOC-amino)ethyl bromide, 96%, Thermo Scientific Chemicals

Catalog number: 398940010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

2-(BOC-amino)ethyl bromide, 96%, Thermo Scientific Chemicals

Catalog number: 398940010
1 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 398940010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
39684-80-5
IUPAC Name
tert-butyl N-(2-bromoethyl)carbamate
Molecular Formula
C7H14BrNO2
InChI Key
TZRQZPMQUXEZMC-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NCCBr
Appearance (Form)
or crystals or powder
Water
=<2 %
Appearance (Form)
Low melting crystalline solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text