Thermo Scientific™

HAAKE™ Viscotester™ 3 Rotational Viscometer

Catalog number: 399-0112
Have Questions?
Thermo Scientific™

HAAKE™ Viscotester™ 3 Rotational Viscometer

Catalog number: 399-0112
Have Questions?

Perform viscosity measurements with ease.  The Thermo Scientific™ HAAKE™ Viscotester™ 3 rotational viscometer ensures quick, reliable tests for QC applications.

Use it as a handheld viscometer or on a lab stand. Color coding ensures measuring geometries match the analysis, the display is readable in any lighting, and the viscometer remembers the reference point measurement for you.  All that comes in a ready-to-go carrying case. 

HAAKE Viscotester 3 Rotational Viscometer is the replacement for the discontinued Thermo Scientific™ HAAKE™ Viscotester™ 1 plus and 2 plus Rotational Viscometers.

 
Catalog Number
399-0112
Unit Size
Each
Description
HAAKE Viscotester 3 Rotational Viscometer for higher viscosity measurements
Viscosity Range
30 to 400 000 mPas
Price (USD)
Full specifications
DescriptionHAAKE Viscotester 3 Rotational Viscometer for higher viscosity measurements
IncludesBase viscometer comes in carrying case with instrument holder (for lab stand), one measuring cup (No. 3), three rotors (No. 1, 2 and 3), four rechargeable batteries and one power supply adapter. Easily upgrade your Thermo Scientific HAAKE Viscotester 2 Plus viscometer to this model; the results are compatible.
Measuring Accuracy± 5% FSD (rotors R1, R2, R3)
± 7.5% FSD (rotor R5)
± 10% FSD (rotor R4)
Power SupplyOperable by battery or power supply
Rotational Speed Range62.5 rpm
Temperature Range (Metric)Up to 150°C
Viscosity Range30 to 400 000 mPas
Unit SizeEach
Catalog Number
Unit Size
Each
Description
HAAKE Viscotester 3 Rotational Viscometer for low viscosity measurements
Viscosity Range
1.5 to 1300 mPas
Price (USD)
Full specifications
DescriptionHAAKE Viscotester 3 Rotational Viscometer for low viscosity measurements
IncludesBase viscometer comes in carrying case with instrument holder (for lab stand), two measuring cups (A and B), three rotors (No. 3, 4 and 5), four rechargeable batteries and one power supply adapter. Easily upgrade your Thermo Scientific HAAKE Viscotester 1 Plus viscometer to this model; the results are compatible.
Measuring Accuracy± 5% FSD (rotors R1, R2, R3)
± 7.5% FSD (rotor R5)
± 10% FSD (rotor R4)
Power SupplyOperable by battery or power supply.
Rotational Speed Range62.5 rpm
Temperature Range (Metric)Up to 150°C
Viscosity Range1.5 to 1300 mPas
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionViscosity RangePrice (USD)
399-0112Full specifications
EachHAAKE Viscotester 3 Rotational Viscometer for higher viscosity measurements30 to 400 000 mPasRequest A Quote
DescriptionHAAKE Viscotester 3 Rotational Viscometer for higher viscosity measurements
IncludesBase viscometer comes in carrying case with instrument holder (for lab stand), one measuring cup (No. 3), three rotors (No. 1, 2 and 3), four rechargeable batteries and one power supply adapter. Easily upgrade your Thermo Scientific HAAKE Viscotester 2 Plus viscometer to this model; the results are compatible.
Measuring Accuracy± 5% FSD (rotors R1, R2, R3)
± 7.5% FSD (rotor R5)
± 10% FSD (rotor R4)
Power SupplyOperable by battery or power supply
Rotational Speed Range62.5 rpm
Temperature Range (Metric)Up to 150°C
Viscosity Range30 to 400 000 mPas
Unit SizeEach
399-0111Full specifications
EachHAAKE Viscotester 3 Rotational Viscometer for low viscosity measurements1.5 to 1300 mPasRequest A Quote
DescriptionHAAKE Viscotester 3 Rotational Viscometer for low viscosity measurements
IncludesBase viscometer comes in carrying case with instrument holder (for lab stand), two measuring cups (A and B), three rotors (No. 3, 4 and 5), four rechargeable batteries and one power supply adapter. Easily upgrade your Thermo Scientific HAAKE Viscotester 1 Plus viscometer to this model; the results are compatible.
Measuring Accuracy± 5% FSD (rotors R1, R2, R3)
± 7.5% FSD (rotor R5)
± 10% FSD (rotor R4)
Power SupplyOperable by battery or power supply.
Rotational Speed Range62.5 rpm
Temperature Range (Metric)Up to 150°C
Viscosity Range1.5 to 1300 mPas
Unit SizeEach
Showing 2 of 2
Designed for simplicity – The HAAKE Viscotester 3 Rotational Viscometer is built for ease of use to get you quickly to the right viscosity measurements:
  • Smart alignment given by design and visualized by an electronic level
  • Quick coupling of measuring geometries for fast and easy adaptation
  • Color coded geometries and a geometry range indicator ensure correct rotor selection every time
  • Adaptable display brightness ensures ideal readability in any environment
  • Battery and power supply options allow the viscometer to be run anywhere
  • Memory assist functionality allows you to quickly and easily compare measured viscosities with the reference point
  • Selectable viscosity units (mPa s or dPa s)
  • Compatibility with previous viscometer models

Recommended for: your entire process, from accepting the delivery of goods, to sampling during production and quality control (QC).
  • Printing inks, paints, inks and other coatings
  • Shampoos, creams, lotions, and similar personal care products
  • Oil, greases, pastes and similar substances
  • Sauces, thickeners, and similar food products

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text