2-Bromo-4,5-difluorobenzoic acid, 97%, Thermo Scientific Chemicals
2-Bromo-4,5-difluorobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-4,5-difluorobenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 399040050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS64695-84-7
IUPAC Name2-bromo-4,5-difluorobenzoic acid
Molecular FormulaC7H3BrF2O2
InChI KeyDGCBGBZYTNTZJH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(F)=C(F)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Melting point118°C to 122°C
HPLC>=96.0 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only