2-Bromo-4,5-difluorobenzoic acid, 97%, Thermo Scientific Chemicals
2-Bromo-4,5-difluorobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-4,5-difluorobenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 399040250
Price (USD)/ Each
234.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
276.00 
Save 41.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
234.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
276.00 
Save 41.35 (15%)
Add to cart
2-Bromo-4,5-difluorobenzoic acid, 97%, Thermo Scientific Chemicals
Catalog number399040250
Price (USD)/ Each
234.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
276.00 
Save 41.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-bromo-4,5-difluorobenzoic acid
Molecular FormulaC7H3BrF2O2
InChI KeyDGCBGBZYTNTZJH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(F)=C(F)C=C1Br
Molecular Weight (g/mol)237.00
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Melting point118°C to 122°C
HPLC>=96.0 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only