Thermo Scientific Chemicals

BOC-S-trityl-L-cysteine, 99%, Thermo Scientific Chemicals

Catalog number: 399060010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

BOC-S-trityl-L-cysteine, 99%, Thermo Scientific Chemicals

Catalog number: 399060010
1 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 399060010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
21947-98-8
IUPAC Name
2-{[(tert-butoxy)carbonyl]amino}-3-[(triphenylmethyl)sulfanyl]propanoic acid
Molecular Formula
C27H29NO4S
InChI Key
JDTOWOURWBDELG-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
HPLC
>=94.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text