4-Thioanisolemagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
4-Thioanisolemagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Thioanisolemagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 399071000
Price (USD)/ Each
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namebromo[4-(methylsulfanyl)phenyl]magnesium
Molecular FormulaC7H7BrMgS
InChI KeySSVQCUSLJVVRCA-UHFFFAOYSA-M
SMILESCSC1=CC=C([Mg]Br)C=C1
Molecular Weight (g/mol)227.40
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay develop some turbidity or precipitate
Appearance (Color)Clear yellow to brown
Appearance (Form)Solution
Titration Grignard0.4 to 0.6 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only