2-(Triphenylphosphoranylidene)propionaldehyde, 98%, Thermo Scientific Chemicals
2-(Triphenylphosphoranylidene)propionaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Triphenylphosphoranylidene)propionaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 399110010
Price (USD)/ Each
39.20
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
39.20
Add to cart
2-(Triphenylphosphoranylidene)propionaldehyde, 98%, Thermo Scientific Chemicals
Catalog number399110010
Price (USD)/ Each
39.20
-
Add to cart
Chemical Identifiers
CAS24720-64-7
IUPAC Name2-(triphenyl-λ⁵-phosphanylidene)propanal
Molecular FormulaC21H19OP
InChI KeyVHUQEFAWBCDBSC-UHFFFAOYSA-N
SMILESCC(C=O)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Powder
Infrared spectrumConforms
NMR (31P-NMR)>=97.5 %
NMR (1H-NMR)>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only