Thermo Scientific Chemicals

2-(Triphenylphosphoranylidene)propionaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 399110100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2-(Triphenylphosphoranylidene)propionaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 399110100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 399110100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
24720-64-7
IUPAC Name
2-(triphenyl-λ⁵-phosphanylidene)propanal
Molecular Formula
C21H19OP
InChI Key
VHUQEFAWBCDBSC-UHFFFAOYSA-N
SMILES
CC(C=O)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to light beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
NMR (31P-NMR)
>=97.5 %
NMR (1H-NMR)
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text