Thermo Scientific Chemicals

5-Formyluracil, 98% , Thermo Scientific Chemicals

Catalog number: 399142500
250 mg, Each
Thermo Scientific Chemicals

5-Formyluracil, 98% , Thermo Scientific Chemicals

Catalog number: 399142500
250 mg, Each
Quantity
Catalog number: 399142500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1195-08-0
IUPAC Name
2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde
Molecular Formula
C5H4N2O3
InChI Key
OHAMXGZMZZWRCA-UHFFFAOYSA-N
SMILES
O=CC1=CNC(=O)NC1=O
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text