Thermo Scientific Chemicals

Ethyl (4-bromobenzoyl)acetate, 95%, Thermo Scientific Chemicals

Catalog number: 399160010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl (4-bromobenzoyl)acetate, 95%, Thermo Scientific Chemicals

Catalog number: 399160010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 399160010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26510-95-2
IUPAC Name
ethyl 3-(4-bromophenyl)-3-oxopropanoate
Molecular Formula
C11H11BrO3
InChI Key
PBDYXCKRDRCJDC-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(=O)C1=CC=C(Br)C=C1
Appearance (Form)
Liquid
Appearance (Color)
Clear yellow
Infrared spectrum
Conforms
HPLC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text