Ethyl (4-bromobenzoyl)acetate, 95%, Thermo Scientific Chemicals
Ethyl (4-bromobenzoyl)acetate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl (4-bromobenzoyl)acetate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 399160050
Price (USD)/ Each
358.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
358.00
Add to cart
Ethyl (4-bromobenzoyl)acetate, 95%, Thermo Scientific Chemicals
Catalog number399160050
Price (USD)/ Each
358.00
-
Add to cart
Chemical Identifiers
IUPAC Nameethyl 3-(4-bromophenyl)-3-oxopropanoate
Molecular FormulaC11H11BrO3
InChI KeyPBDYXCKRDRCJDC-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)C1=CC=C(Br)C=C1
Molecular Weight (g/mol)271.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear yellow
Infrared spectrumConforms
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only