Thermo Scientific Chemicals

Bromomethyl acetate, 95%, Thermo Scientific Chemicals

Catalog number: 399180100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Bromomethyl acetate, 95%, Thermo Scientific Chemicals

Catalog number: 399180100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 399180100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
590-97-6
IUPAC Name
bromomethyl acetate
Molecular Formula
C3H5BrO2
InChI Key
NHYXMAKLBXBVEO-UHFFFAOYSA-N
SMILES
CC(=O)OCBr
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4470 to 1.4540 (20°C, 589 nm)
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text