Thermo Scientific Chemicals

Diethyl 1,4-dihydro-3,5-pyridinecarboxylate, 95%, Thermo Scientific Chemicals

Catalog number: 399190010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl 1,4-dihydro-3,5-pyridinecarboxylate, 95%, Thermo Scientific Chemicals

Catalog number: 399190010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 399190010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1149-23-1
IUPAC Name
3,5-diethyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Formula
C13H19NO4
InChI Key
LJXTYJXBORAIHX-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=C(C)NC(C)=C(C1)C(=O)OCC
Appearance (Color)
Yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure
HPLC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text