Thermo Scientific Chemicals

(R)-(+)-N-BOC-2-Piperidinecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 399250010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(R)-(+)-N-BOC-2-Piperidinecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 399250010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 399250010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
28697-17-8
IUPAC Name
1-[(tert-butoxy)carbonyl]piperidine-2-carboxylic acid
Molecular Formula
C11H19NO4
InChI Key
JQAOHGMPAAWWQO-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)N1CCCCC1C(O)=O
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
120°C to 125°C
Specific optical rotation
+62.0° to +65.0° (20°C, 589 nm) (c=1%, acetic acid)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text