Thermo Scientific Chemicals

Methyl 3-hydroxy-4-methoxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 399260010
1 g, Each
Thermo Scientific Chemicals

Methyl 3-hydroxy-4-methoxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 399260010
1 g, Each
Quantity
Catalog number: 399260010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6702-50-7
IUPAC Name
methyl 3-hydroxy-4-methoxybenzoate
Molecular Formula
C9H10O4
InChI Key
QXOXUEFXRSIYSW-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(O)=C(OC)C=C1
Appearance (Form)
Crystalline powder or solid
Infrared spectrum
Conforms
Melting point
62°C to 67°C
GC
>=97.5 %
Appearance (Color)
Off-white to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text