Thermo Scientific Chemicals

6-Bromooxindole, 97%, Thermo Scientific Chemicals

Catalog number: 399435000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

6-Bromooxindole, 97%, Thermo Scientific Chemicals

Catalog number: 399435000
500 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 399435000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
99365-40-9
IUPAC Name
6-bromo-2,3-dihydro-1H-indol-2-one
Molecular Formula
C8H6BrNO
InChI Key
JARRYVQFBQVOBE-UHFFFAOYSA-N
SMILES
BrC1=CC=C2CC(=O)NC2=C1
Appearance (Form)
Powder
Melting point
210°C to 216°C
Infrared spectrum
Conforms
HPLC
>=96.0 %
Appearance (Color)
White to orange to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text