1-(2-Bromoethyl)naphtalene, 97%, Thermo Scientific Chemicals
1-(2-Bromoethyl)naphtalene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(2-Bromoethyl)naphtalene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 399560050
Price (USD)/ Each
362.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
362.00
Add to cart
1-(2-Bromoethyl)naphtalene, 97%, Thermo Scientific Chemicals
Catalog number399560050
Price (USD)/ Each
362.00
-
Add to cart
Chemical Identifiers
CAS13686-49-2
IUPAC Name1-(2-bromoethyl)naphthalene
Molecular FormulaC12H11Br
InChI KeyGPHCPUFIWQJZOI-UHFFFAOYSA-N
SMILESBrCCC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.6380 to 1.6400 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only