Thermo Scientific Chemicals

(R)-(+)-Propylene oxide, 98+%, Thermo Scientific Chemicals

Catalog number: 399570050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

(R)-(+)-Propylene oxide, 98+%, Thermo Scientific Chemicals

Catalog number: 399570050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 399570050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15448-47-2
IUPAC Name
(2R)-2-methyloxirane
Molecular Formula
C3H6O
InChI Key
GOOHAUXETOMSMM-GSVOUGTGSA-N
SMILES
C[C@@H]1CO1
Appearance (Color)
Colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.3650 to 1.3680 (20°C, 589 nm)
Specific optical rotation
+12° to +15° (20°C, 589 nm) (Neat)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text