2,4,6-Trichloropyridine, 98%, Thermo Scientific Chemicals
2,4,6-Trichloropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Trichloropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 399590010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16063-69-7
IUPAC Name2,4,6-trichloropyridine
Molecular FormulaC5H2Cl3N
InChI KeyFJNNGKMAGDPVIU-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystals or crystalline solid
Infrared spectrumConforms
Melting point31°C to 37°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only