3-Chloro-2,4(3H,5H)-furandione, 97%, Thermo Scientific Chemicals
3-Chloro-2,4(3H,5H)-furandione, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chloro-2,4(3H,5H)-furandione, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 399650500
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
Add to cart
3-Chloro-2,4(3H,5H)-furandione, 97%, Thermo Scientific Chemicals
Catalog number399650500
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Name3-chlorooxolane-2,4-dione
Molecular FormulaC4H3ClO3
InChI KeyDJEJYZZJYYNADA-UHFFFAOYNA-N
SMILESClC1C(=O)COC1=O
Molecular Weight (g/mol)134.52
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
HPLC>=96.0 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only