Thermo Scientific Chemicals

O-(tert-Butyl)hydroxylamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 399680010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

O-(tert-Butyl)hydroxylamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 399680010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 399680010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
39684-28-1
IUPAC Name
hydrogen O-tert-butylhydroxylamine chloride
Molecular Formula
C4H12ClNO
InChI Key
ZBDXGNXNXXPKJI-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC(C)(C)ON
Appearance (Color)
White to light yellow
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Titration Argentometric
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text