Chemical Identifiers
CAS207857-15-6
IUPAC Nametert-butyl N-({[(tert-butoxy)carbonyl]amino}(trifluoromethanesulfonylimino)methyl)carbamate
Molecular FormulaC12H20F3N3O6S
InChI KeyGOQZIPJCBUYLIR-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC(NC(=O)OC(C)(C)C)=NS(=O)(=O)C(F)(F)F
View more