Mono-Ethyl malonate, 95%, Thermo Scientific Chemicals
Mono-Ethyl malonate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mono-Ethyl malonate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
250 g
Packaging:
Glass bottle
Catalog number 399900050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1071-46-1
IUPAC Name3-ethoxy-3-oxopropanoic acid
Molecular FormulaC5H8O4
InChI KeyHGINADPHJQTSKN-UHFFFAOYSA-N
SMILESCCOC(=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Titration with NaOH>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only