4-(Difluoromethoxy)aniline, 98%, Thermo Scientific Chemicals
4-(Difluoromethoxy)aniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Difluoromethoxy)aniline, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 399910050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-(difluoromethoxy)aniline
Molecular FormulaC7H7F2NO
InChI KeyNDEZTSHWEPQVBX-UHFFFAOYSA-N
SMILESNC1=CC=C(OC(F)F)C=C1
Molecular Weight (g/mol)159.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to orange-brown
Refractive index1.5040 to 1.5080 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only