Thermo Scientific Chemicals

4-Amino-2-chloro-5-fluoropyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 399980050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Amino-2-chloro-5-fluoropyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 399980050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 399980050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
155-10-2
IUPAC Name
2-chloro-5-fluoropyrimidin-4-amine
Molecular Formula
C4H3ClFN3
InChI Key
SLQAJWTZUXJPNY-UHFFFAOYSA-N
SMILES
NC1=NC(Cl)=NC=C1F
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure
Melting point
192°C to 196°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text