CryoMed™ Sensors
CryoMed™ Sensors
Thermo Scientific™
CryoMed™ Sensors
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rate freezers with these CryoMed Sensors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
40003851.2/2.0 Cryo Tubes in CryoMed
40003864.0/5.0 Cryo Tubes in CryoMed
40004020.020 Sheath Straw Sample
40004030.062 Sheath Vial Sample
Catalog number 4000385
Price (USD)
-
Request A Quote
For Use With (Equipment):
1.2/2.0 Cryo Tubes in CryoMed
Ask our AI about this Product
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rate freezers with these CryoMed Sensors.
Features:
  • Thermocouple sensors for cryo tubes, sheath straw and vial samples
  • Includes male plug
Recommended for:
Controlled-rate freezing applications
  • Warranty varies by country, please contact us for details
  • Specifications
    DescriptionThermocouple Sensor
    For Use With (Equipment)1.2/2.0 Cryo Tubes in CryoMed
    HoldsCryo Tube, Sheath Straw, Vial
    TypeThermocouple Sensor
    Capacity (English)7.06 cu. ft.
    Capacity (Metric)2 mL
    Plug TypeMale
    Unit SizeEach
    Have questions about this product? Ask our AI assisted search.
    This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
    Documents & Downloads
    Certificates
    Safety Data Sheets
    Share catalog number, name or link