Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with this optional printer.
Have Questions?
Change view
Catalog Number
Type
Voltage
4000565
Thermal Data Printer
120 V
4000566
Thermal Printer Paper
Not Applicable
4000665
Thermal Data Printer
220 V
3 Options
Catalog number 4000565
Price (EUR)
-
Request A Quote
Type:
Thermal Data Printer
Voltage:
120 V
Ask our AI about this Product
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with Thermo Scientific™ Thermal Printers for CryoMed and CryoPlus™ Freezers. These printers are available in 120V or 220V models, and include cable assembly and RS-232 connector kit.
Model 4000565 - 120V, 60Hz printer with cable assembly and RS-232 connector kit
Model 4000665 - 220V, 50Hz printer with cable assembly and RS-232 connector kit
Model 4000566 - Thermal printer paper (5 rolls/package)
Warranty varies by country, please contact us for details
Specifications
For Use With (Equipment)All CryoMed Controlled-Rate Freezers
TypeThermal Data Printer
WarrantyWarranty varies by country, please contact us for details
Voltage120 V
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
0 results displayed, search above for a specific certificate