Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Actual product may vary
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Thermo Scientific™
Racks for the CryoMed™ Controlled-Rate Freezer
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsType
40007083 x 4/5 mL Cryo Racks (Large)Vial Rack Holder
185089Adjustable, 10-positionCanister Rack
400029440 0.25/0.5 mL StrawsStraw Rack
4000303162 Canes, 5 to 6 Ampules/CaneCane Freezing Rack
400031048 Skin Packets (2 level)Skin Rack
400031172 Packets (3 level)Skin Rack
400031224 Packets (1 level)Skin Rack
400034010 Canisters (for cord blood, Model 4000610)Canister Rack
400070077 CanesCane Freezing Rack
400070176 1.2 mL/2 mL AmpulesVial Rack (Small)
40007025 x 1.2/2 mL Cryo Racks (Small)Vial Rack Holder
4000703161 1.2 mL/2 mL AmpulesVial Rack (Large)
40007045 x 1.2/2 mL Cryo Racks (Large)Vial Rack Holder
400070576 4 mL/5 mL AmpulesVial Rack (Small)
40007063 x 4/5 mL Cryo Racks (Small)Vial Rack Holder
4000707161 4 mL/5 mL AmpulesVial Rack (Large)
Catalog number 4000708
Price (EUR)
-
Request A Quote
Holds:
3 x 4/5 mL Cryo Racks (Large)
Type:
Vial Rack Holder
Ask our AI about this Product
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
  • Warranty varies by country, please contact us for details
Specifications
DescriptionThree 4 mL/5 mL Cryo Racks (Large)
For Use With (Equipment)CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458
Holds3 x 4/5 mL Cryo Racks (Large)
ColorSilver
TypeVial Rack Holder
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection