1'-Acetonaphthone, 95%, Thermo Scientific Chemicals
1'-Acetonaphthone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1'-Acetonaphthone, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 400090010
Price (USD)/ Each
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS941-98-0
IUPAC Name1-(naphthalen-1-yl)ethan-1-one
Molecular FormulaC12H10O
InChI KeyQQLIGMASAVJVON-UHFFFAOYSA-N
SMILESCC(=O)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=94 %
Refractive index1.6270 to 1.6290 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only