Thermo Scientific Chemicals

1'-Acetonaphthone, 95%, Thermo Scientific Chemicals

Catalog number: 400092500
250 g, Each
Thermo Scientific Chemicals

1'-Acetonaphthone, 95%, Thermo Scientific Chemicals

Catalog number: 400092500
250 g, Each
Quantity
Catalog number: 400092500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
941-98-0
IUPAC Name
1-(naphthalen-1-yl)ethan-1-one
Molecular Formula
C12H10O
InChI Key
QQLIGMASAVJVON-UHFFFAOYSA-N
SMILES
CC(=O)C1=C2C=CC=CC2=CC=C1
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
GC
>=94 %
Refractive index
1.6270 to 1.6290 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text