Thermo Scientific Chemicals

Acid Fuchsin

Catalog number: 400211000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Acid Fuchsin

Catalog number: 400211000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 400211000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3244-88-0
IUPAC Name
disodium 2-amino-5-[(4-amino-3-sulfophenyl)[(1Z)-4-imino-3-methyl-5-sulfonatocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
Molecular Formula
C20H17N3Na2O9S3
InChI Key
WZRZTHMJPHPAMU-UHFFFAOYSA-L
SMILES
[Na+].[Na+].CC1=C\C(C=C(C1=N)S([O-])(=O)=O)=C(\C1=CC=C(N)C(=C1)S(O)(=O)=O)C1=CC=C(N)C(=C1)S([O-])(=O)=O
Appearance (Color)
Dark green
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
UV Lambda max
540 to 549 nm (10 mg/l in water + 2.5ml 0.1N HCl)
Absorption Ratio P-15/P+15
1.10 to 1.26

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text