Thermo Scientific Chemicals

Acridine-9-carboxylic acid hydrate, 97%, Thermo Scientific Chemicals

Catalog number: 400220050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Acridine-9-carboxylic acid hydrate, 97%, Thermo Scientific Chemicals

Catalog number: 400220050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 400220050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
332927-03-4
IUPAC Name
acridine-9-carboxylic acid
Molecular Formula
C14H9NO2
InChI Key
IYRYQBAAHMBIFT-UHFFFAOYSA-N
SMILES
OC(=O)C1=C2C=CC=CC2=NC2=CC=CC=C12
Water
=<15 % (K.F.)
Appearance (Color)
Bright yellow to yellow-green
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=96.0 % (on dry substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text