Thermo Scientific Chemicals

9(10H)-Acridone, 99%, Thermo Scientific Chemicals

Catalog number: 400250100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

9(10H)-Acridone, 99%, Thermo Scientific Chemicals

Catalog number: 400250100
10 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 400250100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
578-95-0
IUPAC Name
9,10-dihydroacridin-9-one
Molecular Formula
C13H9NO
InChI Key
FZEYVTFCMJSGMP-UHFFFAOYSA-N
SMILES
O=C1C2=CC=CC=C2NC2=CC=CC=C12
Appearance (Color)
Yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with TBAH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text