Bulk CPG (Controlled Pore Glass), 500 Angstrom pore size, T
Thermo Scientific™
Bulk CPG (Controlled Pore Glass), 500 Angstrom pore size, T
Bulk T Controlled Pore Glass (CPG) for users to pack their own columns for synthesis of oligonucleotides • Proprietary designRead more
Have Questions?
Catalog number 400397
Price (USD)/ 1 g
-
Bulk T Controlled Pore Glass (CPG) for users to pack their own columns for synthesis of oligonucleotides <50 bases in length.
• Proprietary design and manufacturing methods ensure consistency and high quality • The 500 angstrom pore size of the support allows synthesis of oligonucleotides of up to 50 bases • Save time and money
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Column TypeOligonucleotide Synthesis Column
FormatBottle
Sample TypeOligos
For Use With (Equipment)3900 DNA Synthesizer
For Use With (Application)Oligo Synthesis
MatrixControlled Pore Glass
Pore Size500 Å
Product TypeOligonucleotide Synthesis Columns
Quantity1 g
Shipping ConditionRoom Temperature
TargetNucleic Acids
Unit Size1 g
Contents & Storage
Store bulk support in a dry environment at room temperature.