Agar, pure, powder
Thermo Scientific Chemicals

Agar, pure, powder

For use in biological culture media.
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
25 kg
500 g
Catalog number 400402500
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
Add to cart
Agar, pure, powder
Catalog number400402500
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9002-18-0
IUPAC Name2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Molecular FormulaC14H24O9
InChI KeyGYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILESCOC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow to beige
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<20.0 % (1 g, 105°C, 5 h)
Heavy metals=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only