Thermo Scientific Chemicals

Agarose medium EEO

Catalog number: 400421000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

Agarose medium EEO

Catalog number: 400421000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 400421000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9012-36-6
IUPAC Name
(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C24H38O19
InChI Key
MJQHZNBUODTQTK-WKGBVCLCSA-N
SMILES
OC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
Infrared spectrum
Conforms
Loss on drying
=<10 % (5 g, 110°C, 24 h)
Melting point
86.5°C to 89.5°C (1.5 % gel)
Sulfate (SO4)
=<0.25 %
Gel strength
>=750 g/cm² (1 % gel)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text