Chemical Identifiers
IUPAC Name(9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride
Molecular FormulaC56H68Cl4CuN16S4
InChI KeyXRWQTDIELZDCPS-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C
Molecular Weight (g/mol)1298.86
View more