Alcian Blue 8GX, certified
Alcian Blue 8GX, certified
Alcian Blue 8GX, certified
Alcian Blue 8GX, certified
Thermo Scientific Chemicals

Alcian Blue 8GX, certified

CAS: 33864-99-2 | C56H68Cl4CuN16S4 | 1298.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 400461000
Price (USD)
988.65
Special offer
Online exclusive
Ends: 26-Sep-2025
1,221.00
Save 232.35 (19%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
988.65
Special offer
Online exclusive
Ends: 26-Sep-2025
1,221.00
Save 232.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS33864-99-2
IUPAC Name(9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride
Molecular FormulaC56H68Cl4CuN16S4
InChI KeyXRWQTDIELZDCPS-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue to purple
Infrared spectrumConforms
Dye content>=50 %
UV Lambda max615 to 635 nm (in water)
BSC CertificationPasses
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link