Thermo Scientific Chemicals

Alizarin Red S, pure, certified

Catalog number: 400480250
25 g, Each
Thermo Scientific Chemicals

Alizarin Red S, pure, certified

Catalog number: 400480250
25 g, Each
Quantity
Catalog number: 400480250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
130-22-3
IUPAC Name
sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Molecular Formula
C14H7NaO7S
InChI Key
HFVAFDPGUJEFBQ-UHFFFAOYSA-M
SMILES
[Na+].OC1=C(O)C2=C(C=C1S([O-])(=O)=O)C(=O)C1=CC=CC=C1C2=O
Appearance (Color)
Yellow to orange to brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
UV Lambda max
554 to 558 nm (in 0.1 M NaOH)
UV Lambda max
590 to 599 nm (in 0.1 M NaOH)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text