Thermo Scientific Chemicals

Aluminum sulfate octadecahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 400590025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Aluminum sulfate octadecahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 400590025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 400590025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-31-8
IUPAC Name
dialuminium(3+) nonahydrate trisulfate
Molecular Formula
Al2H18O21S3
InChI Key
KFCWRPFNNIEZRY-UHFFFAOYSA-H
SMILES
O.O.O.O.O.O.O.O.O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Insoluble matter
=<0.01 %
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals and/or chunks
Titration Complexometric
98.0 to 102.0 %
Chloride (Cl)
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text