Thermo Scientific Chemicals

4-(2-Aminoethyl)morpholine, 98+%, Thermo Scientific Chemicals

Catalog number: 400750050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(2-Aminoethyl)morpholine, 98+%, Thermo Scientific Chemicals

Catalog number: 400750050
5 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 400750050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2038-03-1
IUPAC Name
2-(morpholin-4-yl)ethan-1-amine
Molecular Formula
C6H14N2O
InChI Key
RWIVICVCHVMHMU-UHFFFAOYSA-N
SMILES
NCCN1CCOCC1
Appearance (Color)
Clear colorless to white to yellow
Appearance (Form)
Liquid or solid
Refractive index
1.4740 to 1.4780 (20°C, 589 nm)
GC
>=98.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text