Thermo Scientific Chemicals

8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt hydrate, 80%, tech., Thermo Scientific Chemicals

Catalog number: 400841000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt hydrate, 80%, tech., Thermo Scientific Chemicals

Catalog number: 400841000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 400841000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
343321-59-5
IUPAC Name
sodium 4-amino-5-hydroxy-7-sulfonaphthalene-2-sulfonate hydrate
Molecular Formula
C10H10NNaO8S2
InChI Key
YYIGOAMEITZADH-UHFFFAOYSA-M
SMILES
O.[Na+].NC1=CC(=CC2=CC(=CC(O)=C12)S(O)(=O)=O)S([O-])(=O)=O
Appearance (Color)
White to beige to grey to brown
Appearance (Form)
Powder or crystals and/or lumps
Infrared spectrum
Conforms
HPLC
>=75 %
Loss on drying
=<6 % (1 g, 150°C, 4 h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text