Thermo Scientific Chemicals

4-Aminophthalonitrile, 97%, Thermo Scientific Chemicals

Catalog number: 401020050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Aminophthalonitrile, 97%, Thermo Scientific Chemicals

Catalog number: 401020050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 401020050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
56765-79-8
IUPAC Name
4-aminobenzene-1,2-dicarbonitrile
Molecular Formula
C8H5N3
InChI Key
RRCAJFYQXKPXOJ-UHFFFAOYSA-N
SMILES
NC1=CC=C(C#N)C(=C1)C#N
Appearance (Color)
White to yellow to orange
Appearance (Form)
Crystalline powder or crystals and/or chunks
HPLC
>=96.0 %
Infrared spectrum
Conforms
Melting point
177°C to 182°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text