2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals
2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number 401030050
Price (USD)/ Each
58.40
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
58.40
Add to cart
2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals
Catalog number401030050
Price (USD)/ Each
58.40
-
Add to cart
Chemical Identifiers
CAS534-03-2
IUPAC Name2-aminopropane-1,3-diol
Molecular FormulaC3H9NO2
InChI KeyKJJPLEZQSCZCKE-UHFFFAOYSA-N
SMILESNC(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Adhering crystalline powder
Infrared spectrumConforms
Melting point52°C to 56°C
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only