Ammonium carbonate, ACS reagent
Ammonium carbonate, ACS reagent
Ammonium carbonate, ACS reagent
Ammonium carbonate, ACS reagent
Thermo Scientific Chemicals

Ammonium carbonate, ACS reagent

CAS: 506-87-6 | CH8N2O3 | 96.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
2.5 kg
Packaging:
HDPE plastic bottle
Catalog number 401130010
Price (EUR)
105,40
Online Exclusive
127,00
Save 21,60 (17%)
Each
-
Add to cart
Quantity:
1 kg
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (EUR)
105,40
Online Exclusive
127,00
Save 21,60 (17%)
Each
Add to cart
Chemical Identifiers
CAS506-87-6
IUPAC Namecarbonic acid diamine
Molecular FormulaCH8N2O3
InChI KeyPRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILESN.N.OC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystals and/or chunks
Titration excess HCl>=30.0 % (NH3)
Heavy metals (ICP-OES)=<5 ppm
Non-volatile matter=<0.01 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Label Elements
Signal Word

Warning

Hazard Statements

H302 - Harmful if swallowed

Precautionary Statements

P264 - Wash face, hands and any exposed skin thoroughly after handling

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P312 - Call a POISON CENTER or doctor if you feel unwell

Share catalog number, name or link

1x1 image pixel for data collection